Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICESDP

Catalog No. 11010201 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-010-201 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-010-201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.

Specifications

Color Blue
Product Type Dot label laser sheet
Sterility Non-sterile
For Use With (Application) 0.5 to 1.5mL tubes
Quantity 192 dots/sheet; 20 sheets/Pk.
Diameter (English) 0.37 in.
Diameter (Metric) 9.55 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.