Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 384-Well Skirted PCR Plates

Catalog No. 08408241 Shop All Fisherbrand Products
Change view
Click to view available options
Skirt Style:
Skirted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Skirt Style
08-408-241 Skirted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-408-241
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Offer minimized warp and bowing for better robotic handling

  • Use thin-wall design for maximum heat transfer efficiency
  • Fit most popular 384-well thermal cyclers
  • Specially designed for Applied Biosystems thermal cyclers and DNA sequencers
  • Provide optimized fit for the ABI 3730xl DNA Analyzer
  • Polypropylene
  • Well volume: 30μL

Certifications

Certified free of RNase, DNase, Human DNA, pyrogens, and PCR inhibitors

Compliance

Designed to SBS (Society for Biomolecular Screening) standards

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Free of RNase, DNase, Human DNA, pyrogens and PCR inhibitors, SBS (Society for Biomolecular Screening) standards
Format Standard
Array 16 x 24
Color Yellow
For Use With (Equipment) For ABI 3730xl DNA Analyzer, 384-well thermal cyclers, Applied Biosystems thermal cyclers, DNA sequencers
Skirt Style Skirted
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
For Use With (Application) For better robotic handling, maximum heat transfer efficiency
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.