Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 6 in. (150mm) Ruler
SDP

Catalog No. 09016 Shop All Fisherbrand Products
Catalog No. 09-016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With cork and stopper size guide

  • Blue vinyl
  • Dual-scale ruler; 0.062 in. and 1mm increments
  • Temperature conversion scale on back for –20° to +100°C and –10° to +212°F ranges

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.