Learn More
Description
Compatible with all leading thermal cyclers and MegaBACE™ capillary sequencers†
- Virgin polypropylene
- Clear well bottom for sample visibility
- Highly rigid plate deck
- Sealable using adhesive films and foils, heat seals, and cap strips
- 340μL maximum well volume when used with adhesive and heat seals
- Cut-away corner and alphanumic grid
- Cleanroom produced
Specifications
Specifications
| Array | 8 x 12 |
| Color | Natural |
| Product Type | PCR Microplate |
| Skirt Style | Non-skirt |
| Material | Polypropylene |
| No. of Wells | 96 |
| For Use With (Application) | PCR |
| Rimmed | Yes |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.