Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Acid Etched Microscope Slide

Catalog No. 22161869 Shop All Fisherbrand Products
Change view
Click to view available options
3 x 1 in.; Double-Etched Frosted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-161869 3 x 1 in.; Double-Etched Frosted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-161869

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microscope slide is double-etched frosted

  • Available in pack of 144 and case of 10 packs

Specifications

Frosted Double-Etched Frosted
Type Microscope Slide
Length (English) 3 in.
Quantity 144/Pk.
Width (English) 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.