Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Acrylic Dispenser for Wipes

Catalog No. p-211345 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
06-666-51 Large
06-666-50 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 06-666-51
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acrylic clear holder adds extra weight for one-hand dispensing of tissue wipes.

For use with Kimwipes™

  • Extra weight for one-hand dispensing
  • Clear holder mounts to wall or bench with two strips of double-sided tape or mounting screws (not included)
  • Sized for small or large Kimwipes boxes

Specifications

Product Type Wiper Dispenser
Color Clear
Material Acrylic
Mounting Free-standing or Wall Mount
For Use With (Equipment) For Kimwipes™
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.