Learn More
Liberty Industries Adhesive Roll Mop
SDP
Catalog No. 19168113 Shop All Liberty Industries Products
Change view
Click to view available options
Product Type:
4 Rolls and Extendable Handle
Refill Roll
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-168-113 Refill Roll
19-168-112 4 Rolls and Extendable Handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-168-113 Supplier Liberty Industries Supplier No. 10 1127
Add to Cart
Add to Cart
Effectively removes dirt, debris and lint
  • Suitable for use on many surfaces including glass, plastic, metal and cloth
  • Picks up contaminants from hard-to-reach areas such as walls, ceilings, floors, benchtops, windows and corners
  • Rolling mop unit with adhesive sheets connects to lightweight, 4 ft. long (1.2m) aluminum handle—handle extends to 7 ft. (2.1m)
  • Roll is 18 in. wide (46cm) and contains 20 disposable, high-tack polyfoam sheets with bounce-back memory
  • Perforations approximately every 14 in. (41cm) give easy access to fresh sheet
Specifications
Product Type Refill Roll
Width (English) 18 in.
Includes Roll Contains 20 Disposable, High-tack Polyfoam Sheets
For Use With (Equipment) Adhesive Roll Mop, use on Glass, Plastic, Metal and Cloth in hard-to-reach Places
For Use With (Application) Effectively Removes Dirt, Debris and Lint