Learn More
Fisherbrand™ Alkacid™ Full-Range pH Kit
Catalog No. A981 Shop All Fisherbrand Products
Change view
Click to view available options
Range: 0.0 to 3.0; Blue
Catalog No.
A981 Range: 0.0 to 3.0; Blue
1 options
Catalog No. A981
pH accuracy to ±0.25
Provides sharp color change for each 0.5pH unit. Consists of A979 wide-range paper for determining approximate pH value and six specific short-range papers.
  • Range 1 (pH 0.0 to 3.0)
  • Range 2 (pH 3.5 to 5.5)
  • Range 3 (pH 6.0 to 8.5)
  • Range 4 (pH 9.0 to 11.0)
  • Range 5 (pH 10.5 to 12.5)
  • Range 6 (pH 12.0 to 14.0)
Order Info
Each paper comes in 15 ft.L x 0.25 in. W (4.6m x 6.4mm) roll with its own plastic dispenser. Appropriate color chart mounted inside. Seven rolls packed in hinged plastic box. Replacement short-range paper rolls available separately.