Learn More
Raidians Bacteriostatic Additive
SDP
For use with 6 to 20 gal. portable eyewash units
Supplier:  Raidians 01200ST
Catalog No. 19-041-537
Only null left
Add to Cart
Description
Description
Mix one bottle with 6 to 20 gal. (22.7 to 75.7L) of potable water to prevent bacteria, fungi, and algae growth.
Product Suggestions
Product Suggestions