Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Secure Click™ Respirator N95 Particulate Filter

Catalog No. 17100835 Shop All 3M Products
Change view
Click to view available options
Quantity:
200 Filters/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-100-835 200 Filters/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Not for use in environments that are immediately dangerous to life or health (IDLH), sandblasting, or in environments that contain asbestos.
Catalog No. 17-100-835 Supplier 3M™ Supplier No. 7100213450/DEL
Not for use in environments that are immediately dangerous to life or health (IDLH), sandblasting, or in environments that contain asbestos.

This Secure Click™ Particulate Filter N95 can be attached to 3M™ Secure Click™ Cartridges D8000 Series to provide particulate protection when used with 3M™ Secure Click™ facepieces.

  • Works with 3M™ Secure Click™ Cartridges D8000 Series
  • Held in place by required 3M™ Secure Click™ Filter Retainer D701
  • NIOSH-approved N95 particulate filter for respiratory protection against certain non-oil based particulates
  • Allows for replacement of filters prior to change out of cartridges for cost savings
  • Use two D7N11 filters per D8000 Series cartridge for a total of 4 filters per facepiece
  • For workplace/occupational use only
  • Common uses include filtering solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances
  • May also be used for liquid or oil based particles from sprays that do not also emit harmful vapors and for metal fumes produced from welding, brazing, cutting and other operations involving heating of metals
  • Applications: Grinding
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Particulate
Certifications/Compliance NIOSH Approved
Connection Type Bayonet
Color White
Design Type Secure Click
Filter Type N
Product Type Filter
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Equipment) Secure Click Half Facepiece HF-800 Reusable Respirators
Model D7000
Quantity 200 Filters/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.