Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Beta Storage Box

Catalog No. 1200913 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
18cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
12-009-13 18cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-009-13

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Holds two tube holders side-by-side

  • Safely stores reagent bottles containing beta-emitting isotopes
  • Made of 0.375 in. thick (0.95cm) clear acrylic with nonskid feet
  • Cover has handle and removes easily
  • Dimensions: 9L x 6W x 7 in.H (23 x 15 x 18cm)

Specifications

Product Type Beta storage box
Color Clear
Includes Lid and radiation warning labels, non-skid feet
Thickness (English) 0.37 in.
Length (Metric) 23cm
Width (Metric) 15cm
Height (English) 7 in.
Material Acrylic
Width (English) 6 in.
Dimensions (L x W x H) 9 x 6 x 7 in. (23 x 15 x 18cm)
For Use With (Application) To store various racks, reagent bottles, and other vessels containing beta-emitting isotopes
Holds 2 Tubes side-by-side
Length (English) 9 in.
Height (Metric) 18cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.