Learn More
Fisherbrand™ Biohazard Mailers
Catalog No. 035237 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Double-mailing Containers
Dimensions (L x W):
2.18 x 5.75 in.
Catalog No. Type Dimensions (L x W)
03-523-7 Double-mailing Containers 2.18 x 5.75 in.
1 options
Catalog No. 03-523-7
Double-mailing containers with inner aluminum screw-cap receptacle and outer fiberboard screw-cap mailing case
  • Both caps have type SC safety edges
  • Ideal for etiologic agents
  • Biohazard specimens
  • TB and HTLV-3 confirmatory test samples
  • Laboratory and slide specimens, reagents and chemicals, glassware and bottles, urine specimen collection
Compliance
Meet static pressure test 95kPa. Conform with the Interstate Quarantine Regulations for Etiologic Agents (42CFR, Part 72.25) when appropriate labeling and absorbent packing materials are used.
Specifications
Type Double-mailing Containers
Dimensions (L x W) 2.18 x 5.75 in.