Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Biohazard Specimen Bags

Catalog No. p-32584 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
9 x 5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-075-388E 9 x 5 in.
19-075-388F -
19-075-388D -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-075-388E
Only null left
Add to Cart
Edge
Add to Cart

Access pocket without opening sealed pouch

  • Two-pouch bags constructed of polyethylene
  • 2mil-thick zipper-seal pouch holds specimen container
  • Separate 1.5mil-thick pocket for papers Each bag printed with biohazard graphics in orange and blue

Specifications

Certifications/Compliance FDA
Color White
For Use With (Application) Access pocket without opening sealed pouch
Includes Printed with biohazard graphics, zipper closure, 2mil thick pouch, 1.5mil thick pocket
Material Polyethylene
Thickness (English) 2 mil
Dimensions (L x W) 9 x 5 in.
Length (English) 9 in.
Width (English) 5 in.
Product Type Biohazard Specimen Bag
Length (Metric) 23 cm
Width (Metric) 13 cm
Thickness (Metric) 0.279 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.