Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bottle Rack

Catalog No. 02993 Shop All Fisherbrand Products
Change view
Click to view available options
Positions:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
02-993 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-993
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wood bottle rack

  • Holds six 30mL round dropping bottles
  • Oil treated wood block with rubber feet, drilled with holes 1.5 in. diameter
  • Size: 12W x 2.25 in. D

Specifications

Positions 6
Product Type Bottle Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.