Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Caution: Contains Formaldehyde Labels
GSA_VAGREENER_CHOICESDP

Catalog No. 18999929 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
320/Pk.
640/Pk.
Size (English):
0.5 x 1.5 in.
0.87 x 3 in.
Size (Metric):
1.27 x 3.81 cm
2.2 x 7.62 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size (English) Size (Metric)
18-999-929 640/Pk. 0.5 x 1.5 in. 1.27 x 3.81 cm
18-999-928 320/Pk. 0.87 x 3 in. 2.2 x 7.62 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-929
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

Strong adhesive ensures that labels remain in place.

Compliance

Meet OSHA requirements.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Color White, Black, Red
Format Label
Material Paper
Product Type Biohazard Warning Tape
Display Text Contains Formaldehylde
For Use With (Application) Biohazard
Quantity 640/Pk.
Size (English) 0.5 x 1.5 in.
Size (Metric) 1.27 x 3.81 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.