Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cheesecloth Wipers

Catalog No. 0666529 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
18 x 36 in. (45.7 x 91.4 cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
06-665-29 18 x 36 in. (45.7 x 91.4 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-665-29

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent-grade quality

  • 100% brilliant white, pure cotton construction will not scratch sensitive surfaces Have a high degree of cleanliness, softness, absorbency and strength
  • Can be used to safely clean microscope and camera optical parts, as well as delicate glassware, syringes, pipets, and pH electrodes
  • Absorb six times their weight in liquids and are useful for mopping up solvents and aqueous solutions
  • Inert to even harsh solvents; ideal as filtering material
  • Grade 10 (mesh size)

Order Info

Each wipe is 18 x 36 in. (46 x 91cm). Sixty wipes per dispenser box (two boxes per pack). Box dimensions: 9 1/2L x 9 1/2W x 4 1/2 in.H (24 x 24 x 11cm).

Compliance

Meet USP and Federal Specification CCC-G-101c.

Specifications

Product Type Cheesecloth Wipe
Length (English) 18 in.
Length (Metric) 45.7 cm
Width (English) 36 in.
Width (Metric) 91.4 cm
Material Cotton
Certifications/Compliance USP, CCC-G-101c
Color White
Dimensions (L x W) 18 x 36 in. (45.7 x 91.4 cm)
For Use With (Application) For use to wipe cuvettes and microscopes, clean cameras, and dry delicate Glassware, syringes, pipettes, and pH electrode
For Use With (Equipment) Microscopes, optical surfaces, Glassware, pH electrodes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.