Description
- Polycarbonate, high-visibility lens absorbs >99.9% of harmful UV light and includes a permanent antifog coating
- Wraparound frame with sideshields and browguard
- Adjustable temple lengths
- Lightweight
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | Meets ANSI Z87.1-2003 standards. |
| Lens Tint | Clear |
| Frame Color | Black |
| Temple Style | Adjustable length |
| Lens Material | Polycarbonate |