Learn More
Fisherbrand™ Contempo Safety Glasses
Catalog No. 191302094 Shop All Fisherbrand Products
Change view
Click to view available options
Frame Color:
Black
Blue
Lens Tint:
Clear
Tinted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-130-2094 Blue Clear
19-130-2092 Black Clear
19-130-2093 Black Tinted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-2094
Add to Cart
Add to Cart
This item is not returnable. View return policy
Offer single-lens styling with a dual-lens look
  • Polycarbonate, high-visibility lens absorbs >99.9% of harmful UV light and includes a permanent antifog coating
  • Wraparound frame with sideshields and browguard
  • Adjustable temple lengths
  • Lightweight
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating Anti-fog
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Lens Tint Clear
Frame Color Blue
Temple Style Adjustable length
Lens Material Polycarbonate