Learn More
Description
- Choice of two grades: laboratory grade (corks cut from selected wood), or superior grade (corks cut from high-quality wood)
- Assorted lengths; Sizes: 6-15
Specifications
Specifications
| Product Type | Cork Stopper |
| Stopper Number | Assorted |
| Size | 6 to 15 |
| Grade | Superior |
| Material | Wood |
| For Use With (Application) | Ideal for sealing bottles |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.