Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cover for Biohazard Incinerator Disposal Carton

Catalog No. 03421525 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-421-525 White, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-421-525
  • $70.25
    $52.65 / Each

    Save $17.60

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $452.00
    $338.65 / Case of 8 EA

    Save $113.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Opens with a single touch.

  • Dishwasher-safe and autoclave-safe, the reusable disposal carton cover opens quickly and easily by pressing the built-in release tab, no batteries required
  • Store the disposable cardboard lid on the bottom of the carton and when carton is full, simply remove the reusable Disposal Carton Cover, replace with the cardboard lid, and dispose of the carton in the usual fashion
  • Autoclavable at 250°F (121°C)
  • Fits any disposal carton that has a 12 x 12 in. (30.5 x 30.5 cm) opening including Fisherbrand Burn-up Bin Biohazard Waste Boxes 12-009-8A and 19-168-430
  • Has a smooth 4 x 4 in. (10.1 x 10.1 cm) area on top for applying labels
  • Provides a wider opening 20.5 x 24.4cm (8 x 9.5 in.) permitting safer and more sanitary deposit of wastes
  • Center panel is red to match the printing on the Biohazard disposal cartons providing fast visual recognition of the waste stream
  • Wash as needed

Specifications

Color White, Red
Autoclavable Autoclavable
For Use With (Equipment) For disposal cartons with 12 x 12 in. opening
Product Type Carton Cover
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.