Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Custom Cleaned Clear Boston Rounds
GREENER_CHOICE

Catalog No. 02912011 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-912-011 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-912-011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Containers and caps are noncertified and precleaned with USP purified water

  • With attached PTFE-faced, PE-lined caps
  • Containers and caps are noncertified and precleaned with USP Purified Water for visible particulates in a clean process area

Order Info

Custom packaging and certification available upon request

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Cleaning Procedure USP purified Water
Color Clear
Material Glass
Length (Metric) 63mm
Quantity 24/PK.
Closure Color White
Liner Material PTFE
Closure Included Yes
Mouth Wide
Closure Liner Polyethylene
Closure Material PTFE
Closure Size 22-400 mm
Closure Type Screw Cap
Shape Round
Product Type Boston Round Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.