Learn More
Fisherbrand™ Deuterium Lamps for Background Correction
Catalog No. 1438625L Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
AA Spectrometer
Spectrometer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-386-25L Spectrometer
14-386-25R AA Spectrometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning:for safe operation of deuterium lamps, the operator should never look directly at the lamp. Products produce high levels of deep UV light, and it is imperative that appropriate eye protection is used. Also, the envelope can reach 200°C during operation, and proper precautions must be taken if lamp is to be handled.
Catalog No. 14-386-25L
Warning:for safe operation of deuterium lamps, the operator should never look directly at the lamp. Products produce high levels of deep UV light, and it is imperative that appropriate eye protection is used. Also, the envelope can reach 200°C during operation, and proper precautions must be taken if lamp is to be handled.