Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Diagnostic Foam Rack Kit

Catalog No. 19075386
Change view
Click to view available options
Capacity:
Holds 45 tubes
Dimensions (D x W x H) Interior:
4 x 9 x 5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Dimensions (D x W x H) Interior
19-075-386 Holds 45 tubes 4 x 9 x 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-075-386 Supplier Fisherbrand™ Supplier No. FSB2200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

One Pick-n-Pluck™ foam block; One 14W x 12 in. H (35.6 x 30.5cm) Bitran bag with reclosable, double-locking, liquid-tight zipper; Attached document pouch and biohazard symbol; Two 250mL absorbent sheets

Convenient kit includes everything needed to transport tubes safely and efficiently

  • Foam block holds up to 45 tubes
  • Liquid-tight outer bag with reclosable zipper surrounds foam and tubes
  • Two absorbent sheets contain potential spills
  • Requisition pouch keeps paperwork dry and handy
  • OSHA-required biohazard symbol included

  • Unit Dimensions: 9 × 5 × 4 in. (22.9 × 12.7 × 10.2cm) Block

Specifications

Dimensions (D x W x H) Interior 4 x 9 x 5 in.
Capacity Holds 45 tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.