Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Diagnostic Mailer

Catalog No. 19038298 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H) Exterior:
23.5 x 11.4 x 5.7 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Exterior
19-038-298 23.5 x 11.4 x 5.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-038-298
Add to Cart
Add to Cart

Includes

One 9 x 12 in. (22.9 x 30.5cm) liquid-tight Infecon bag with document pouch and biohazard symbol, one 6-tube bubble pouch, one 250mL capacity absorbent sheet, one shipping box

For secure shipping of diagnostic specimens

  • Quantity per case: 36
  • Unit Dimensions: 9.25 × 4.5 × 2.25 in. (23.5 × 11.4 × 5.7cm)

Specifications

Applications Secure shipping of diagnostic specimens
Dimensions (L x W x H) Exterior 23.5 x 11.4 x 5.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.