Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Dispette™ 2 Disposable ESR kit (Pipettes and Reservoirs)

Catalog No. 02675179 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-675-179 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-675-179

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

100 triple-plugged Dispette™ Pipettes & 100 reservoirs pre-filled with 0.25mL of 0.9% Saline

Provides convenient and safe ESR determinations through a technique correlating directly with the Westergren method

Manufactured with a minimum 2.55mm internal bore to prevent “cell bouncing” or the impedance of proper cell sedimentation.

  • Polystyrene design is shatterproof and safe to handle
  • “Triple plug” tip design provides a unique protective barrier from accidental overfilling of the pipet tube and spillage of blood
  • Pierceable reservoir cap provides greater tactile feedback during pipet insertion and eliminates the need to remove the cap before insertion, improving safety
  • Reservoir cap provides enlarged mixing space to ensure adequate mixing of blood and saline
  • “Sealing ring” molded into reservoir below fill line ensures correct filling when inserted into the reservoir and helps to prevent leaks
  • Restricted end of reservoir acts as an extra safety feature — slows downward movement of the tube during insertion and providing a buffered stop
  • Pre-filled reservoirs ensure the correct dilution following recommended Westergren ESR method when 1mL of EDTA blood is added

Compliance

CLIA-waived. Meets NCCLS/ICSH guidelines

Specifications

Certifications/Compliance CLSI, H02-A5, NCCLS/ICSH
Content And Storage 10°C to 24°C
Clia Complexity Waived
Type Dispette 2; Saline ESR Kit
Disposable Single-use
For Use With (Application) For qualitative estimation of the erythrocyte sedimentation rate (ESR or Sed-Rate)
Includes 100 Plugged Dispette; 2 pipets, 100 filling reservoirs containing 0.25mL Normal Saline diluent, triple plug tip, pierceable reservoir cap, sealing ring
Material Polystyrene
Methods Compatibility Westergren Method
Model Dispette 2
Quantity Each
Show More Show Less

DISPETTE is a trademark of Guest Scientific AG, Switzerland

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.