Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Controlled-Drop Pasteur Pipets
SDP

Catalog No. 136786C Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Borosilicate Glass
Flint Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
13-678-6C Flint Glass
13-678-30 Borosilicate Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-678-6C

Perfect for blood typing and cross-matching procedures in blood bank laboratories

  • Deliver 22 to 26 drops per mL
  • Correspond to drop size from antisera reagent bottle droppers
  • Can be cotton plugged (plugs not included)

Order Info

250 Pipets per pack, four packs per case.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner Tip Opening: 1.20 to 1.65 mm
Diameter (Metric) Outer 6.95 mm (top)
Material Flint Glass
For Use With (Application) For blood typing and cross-matching procedures in blood bank laboratories
Length (English) 5.75 in.
Volume (Metric) 2 mL
Length (Metric) 14.6 cm
Type Controlled Drop Pasteur Pipet
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.