Learn More
Fisherbrand™ Disposable Face Shields
Catalog No. 19181600B Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
19-181-600B Medium
1 options
This item is not returnable. View return policy
Catalog No. 19-181-600B
This item is not returnable. View return policy
High-quality, optically clear, distortion-free 7mil film
  • Polyester lens
  • Treated with antifog and antistatic coating
  • Headpiece is held in place by a stretch strap
  • Includes a latex-free foam band for comfortable extended wear
Specifications
Width (Metric) 30.6 cm
Height (Metric) 14.4 cm
Lens Coating Anti-fog, Anti-static
Product Type Faceshield
For Use With (Application) Provides excellent visibility while protecting the face and neck from accidental splashes
Headband Material Elastic Knitted Polyurethane
No. per Pack 25/Pk.
Size Medium
Material PET