Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Canvas Gloves with PVC Dot Grip

Catalog No. 19049583 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-049-583 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-049-583
  • $40.25
    $27.65 / Dozen of 12

    Save $12.60

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $875.00
    $601.65 / Case of 25 DZ

    Save $273.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for light protection in oil drilling, warehousing, farming and automotive applications. PVC dots on the palm and pointer finger help to enhance grip capabilities.

  • Knit wrist prevents dirt and debris from entering the glove
  • PVC dots enhance grip capabilities

Specifications

Color Natural
Grade Economy
Hand Specific
Material Cotton
Product Type General Purpose Gloves
Size Small
Style Clute
Gender Men's
Glove Cuff Style Knit
Thumb Style Straight
Yarn Weight 8 oz.
Length (English) 10.8 in.
Length (Metric) 27.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.