Learn More
Fisherbrand™ Dust'it™ Air Duster
Catalog No. 1464959 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
56.7 g
Catalog No. Volume (Metric)
14-649-59 56.7 g
1 options
This item is not returnable. View return policy
Catalog No. 14-649-59
  • $110.60
    $93.65 / Pack of 3
    Save $16.95
    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Powerful bulb duster forcibly blows away dust, lint and dirt
Ideal for optical lenses, slides, circuit boards and cuvets.
  • Provides high-pressure, pinpoint jets of air exactly where desired; vary squeeze to control delivery
  • One-way inlet valve at top stops air from being drawn through tip
  • Nonconductive plastic sleeve-tip prevents scratches
Specifications
Product Type Air Duster
Bristle Material None
Bristle Type None
Color-coded No
Form Aerosol
Handle Type None
Scent Unscented
Sterility Non-sterile
For Use With (Application) Eliminates dust, lint and dirt; cleans optical lenses, slides and other delicate, difficult-to-clean and inaccessible equipment
Volume (English) 2 oz.
Volume (Metric) 56.7 g