Learn More
Fisherbrand™ EndoCervex™-Brush-S
Catalog No. 14560101 Shop All Fisherbrand Products
Change view
Click to view available options
Style:
Small
Catalog No. Style
14-560-101 Small
1 options
This item is not returnable. View return policy
Catalog No. 14-560-101
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Smaller brush head for pap smear collection
  • Designed specifically for teens, young nulliparous women, or elderly women with a retracted transformation zone and/or stenotic entry of the endocervical canal
  • Stiff, yet flexible head has over 200 hairs for simultaneous collection of cellular material
  • Safety ring below the head decreases the risk of endometrium penetration during collection
  • Head can be detached from handle after specimen collection
  • Suitable for use with conventional cytology and liquid-based cytology collection and transport processes
  • Enhances specimen-to-slide transfer
Specifications
Style Small