Learn More
Fisherbrand™ EyeSafe™ Faucet-Mounted Eyewash
Catalog No. 19050632
Click to view available options
Mounting:
Faucet
Catalog No. Mounting
19-050-632 Faucet
1 options
Catalog No. 19-050-632 Supplier Fisherbrand™ Supplier No. G1100
Includes
Float-off dust covers to protect outlet heads and three adaptors (0.94 in. -27, 0.81 in. -27 and 0.38 in. IPS) for installation on most commonly used faucets.
Converts any faucet into an emergency eye wash station
  • Does not interfere with normal faucet operation
  • Can be installed at any sink, close to where accidents might occur, to provide an unlimited supply of potable water for rinsing the user's eyes
  • Outlet heads are mounted 3 in. apart and deliver a soft, aerated flow of water
  • Heads angle forward toward user and angle is adjustable to permit full coverage and avoid splashing
  • Forged brass diverter valve activates with pull knob
  • Inlet body has 0.86 in.-27 female thread
Order Info
Each unit is completely assembled and water tested prior to shipment.
Specifications
For Use With (Application) Converts any Faucet to an Emergency Eyewash Station
Includes Float-off dust covers; Aerator on outlets; Three adapters (0.94 in. -27, 0.81 in. -27 and 0.38 in. IPS) for installation on most commonly used faucets
Mounting Faucet
Product Type Faucet Mounted Eyewash