Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ FE150 Benchtop pH Meter Education Set
SCI_ED

Catalog No. S35923 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
pH Meter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S35923 pH Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S35923
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Measure the acidity or alkalinity of a sample to a high degree of accuracy with the Fisherbrand™ FE150. Its intuitive, simple operation, easy-to-use interface and high accuracy measurements ensure maximum productivity in your classroom or lab.

Comes complete with pH Meter, Single Junction Gel pH Electrode, ATC Probe, and Stand.
TRUSTED_SUSTAINABILITY

Specifications

Includes FE150 pH Meter, Single Junction Gel pH Electrode, ATC Probe, Stand
For Use With (Application) Education
Measurement Modes pH, mV, Relative mV, Temperature
Calibration pH, ORP, Temperature
Range (pH) 0.00 to 14.00
Resolution (pH) 0.01
Range (mV) -1999 mV to 1999 mV
Calibration Points 1 to 5
Temperature Range (Metric) 0°C to 100°C
Display Type LCD
Backlight Option None
Data Points 100
Log Function Type Automatic, Manual
Model FE150 Benchtop pH Meter Education Set
Type pH Meter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.