Description
White cards are made from a blend of 100% cotton and cellulose fibers
- Excellent absorbent padding for the collection of excess reagents or wash solution
- Cellulose fibers improve the card's strength and durability during centrifugation and facilitate the removal of the filter card from the slide
Specifications
Specifications
| For Use With (Equipment) | Shandon Cytospin |
| Type | Filter Card |