Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Kjeldahl Flask Carrier

Catalog No. 1381450 Shop All Fisherbrand Products
Change view
Click to view available options
Positions:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
13-814-50 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-814-50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fisherbrand Kjeldahl Flask Carrier can hold up to six flasks

  • Fisherbrand™ Kjeldahl Flask Carrier can hold up to six flasks (800ml)
  • Spring clamp locking system holds flask necks securely to prevent breakage
  • Holds flasks of sizes: 300 mL, 500 mL, 650 mL or 800 mL
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines

Specifications

Autoclavable Not Autoclavable
Holds Kjeldahl Flasks
Positions 6
Size 300, 500, 650 or 800 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.