Learn More
Fisher Science Education™ Flowering Plants Angiospermae Basic Slide Set
SCI_ED
Catalog No. S68785 Shop All Fisherbrand Products
Change view
Click to view available options
Flowering Plants Slide Set
Catalog No.
S68785 Flowering Plants Slide Set
1 options
Catalog No. S68785
Variety of flowering plants.
Supplied with plastic box for 25 microscope slides.
Set of six slides includes:
  • Mitosis, l.s. from Allium root tips showing all stages of plant mitosis carefully stained with iron-hematoxyline after Heidenhain
  • Monocot and dicot roots, two t.s. on one slide for comparison
  • Monocot and dicot stems, two t.s. on one slide for comparison of the different structures
  • Syringa, lilac, t.s. of a typical mesophytic dicot leaf for general study, showing all structures very clearly
  • Lilium, anther t.s. for general study showing pollen chambers and pollen grains
  • Lilium, ovary t.s., showing arrangement of ovules and all structures for general study
Specifications
Type Flowering Plants Slide Set