Learn More
Fisherbrand™ Fluorescence Rectangular Cells: Cell for Magic Stirrer-Semi-Micro With PTFE Lids, 4 Clear Windows
Shop All Fisherbrand ProductsDescription
Includes
PTFE lid or stopper
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Specifications
| Type | Semi-micro Fluorescence Cell |
| Capacity (Metric) | 1500 μL |
| Material | Quartz |
| Closure Material | PTFE |
| Width (Metric) Interior | 4mm |
| Quantity | Ea., 4 Ea./Cs. |
| Path Length | 10 mm |
| Wavelength Range | 200 to 2500 nm |
| Height (Metric) | 49.5 mm |
| For Use With (Equipment) | Standard Spectrophotometer Cell Holder |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.