Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Glucose Tolerance Beverages

Catalog No. tg30006p Shop All Fisherbrand Products
Change view
Click to view available options
:
Dexicola; 100g
Dexicola; 50g
LIMEONDEX; 50g
SUN-DEX; 100g
SUN-DEX; 50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
TG30006P SUN-DEX; 100g
TG30009P SUN-DEX; 50g
TG86150P Dexicola; 50g
TG86350P LIMEONDEX; 50g
TG30008P Dexicola; 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. TG30006P
This item is not returnable. View return policy

New all-plastic bottling

  • Choice of 50-, 75-, or 100-gram dose per 10-oz. bottle
  • Flavors include SUN-DEX™ (orange), LIMEONDEX (lemon-lime), or DEXICOLA™ (cola) (caffeine-free)
  • Carbonated
  • Meet FDA standards and are Kosher-certified
  • Plastic, tamper-evident caps
  • Volume graduations clearly printed on labels
  • Packaged 24 × 10 fl. oz./Cs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.