Learn More
Fisherbrand™ Gram✓™ Control/Test Slides
GSA_VA
Catalog No. 08801 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
08-801 50/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 08-801
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gram Check Test Slides are provided with known gram positive and gram negative organism control blocks and six additional test blocks for your unknown specimens.
The Gram Check slide is an eight-grid slide with six blank grids. Two of the eight grids have fixed smears of Gram-positive S. aureus and Gram-negative E. coli as Quality Control Grids to provide positive identifications of organisms after gram staining procedure has been conducted.
Order Info
Packaged in individual foil envelopes with instructions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 50/Pk.