Description
Includes
Foamed PE-lined caps
- Narrow mouth is ideal for liquids
- Available as round- or square-shaped
Specifications
Specifications
| Capacity (Metric) | 3785 mL |
| Material | HDPE |
| Mouth | Narrow |
| Closure Liner | Polyethylene Foam |
| Closure Material | Polyethylene |
| Closure Size | 38-400 mm |
| Closure Type | Screw Cap |
| Shape | Square |