Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 18 mm Headpace Magnetic Screw Cap with Septa, 8 mm Hole
GSA_VA

Catalog No. 13622136 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-622-136 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-622-136
Get an Extra Case of Product with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use Fisherbrand™ magnetic headspace vial seals with PAL/CTC/Leap Technologies, Gerstel and other magnetic transport autosamplers.

  • Use magnetic seals with PAL/CTC/Leap Technologies, Gerstel and other magnetic transport autosamplers
  • Caps only work on Precision Thread Vials not other 18 mm screw thread vials

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Material Steel
Pre-slit No
Septum Blue PTFE/White Silicone
No. per Pack 100
Hardness 55 Shore A
Product Line Fisherbrand
Thickness (Metric) 1.5 mm
Closure Size 18 mm
Diameter (Metric) Hole 8 mm
Product Type Headspace Vial Magnetic/Screw Thread Closure
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.