Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Heavy-Paper Lined Sample Bags

Ideal for collection, shipment or storage of grain, seed, soil, or food samples

 View more versions of this product

Catalog No. 01-818-11


Add to cart

Description

Description

Ideal for collection, shipment or storage of grain, seed, soil, or food samples

  • Heavy-paper bags lined with 1mil-thick polyethylene
  • Heat sealed at bottom to prevent seepage
  • Tin-tie closure ensures moisture retention

Order Info

Maximum gusset width on side of bag is 2.5 in. (6.4cm) for 01-818-10; 3 in. (7.6cm) for 01-818-11.

Specifications

Specifications

13cm
5 in.
2 lb.
Assorted
Paper
3 in.
Poly lined paper with tin tie
500
1 mil
No
.9kg
23cm
9 x 5 in. (23 x 13cm)
7.6cm
9 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit