Learn More
Fisherbrand™ Histosette™ II Biopsy Processing/Embedding Cassettes with Separate Lid and Base
Catalog No. 15182709F Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Gray
Green
Orange
Peach
Pink
Tan
White
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-182-709F Aqua
15-182-709G Gray
15-182-709H Orange
15-182-709I Peach
15-182-709K Tan
15-182-709A White
15-182-709B Pink
15-182-709C Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 15-182-709F
Add to Cart
Add to Cart
This item is not returnable. View return policy
Histosette™ II Biopsy Processing/Embedding Cassettes with Separate Lid and Base open and close as often as necessary - cassettes will always relock securely.
  • Disposable, plastic cassettes hold biopsy specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Pore Style Slots
Writing Face Angle 45°
Closure Type Separate Lid
Color Aqua
Material Plastic
Product Type Biopsy Cassette