Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ HMIG and NFR Write-On Labels
GREENER_CHOICESDPSCI_ED

Catalog No. S98411 Shop All Fisherbrand Products
Change view
Click to view available options
Display Text:
HMIG
NFR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
S98411 NFR
S98410 HMIG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S98411
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers.

  • HMIG paper labels are large enough to include the chemical name, common name and manufacturer name
  • NFR labels let you write in the chemical name, owner, date and hazard rating

Order Info

Labels measure 3.87L x 1.5 in. W (9.8 x 3.8cm).

Specifications

Format Label
Product Type NFR Write-On™ Label
Display Text NFR
For Use With (Application) Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers
Quantity 1000/Pk.
Length (English) 3.87 in.
Length (Metric) 9.8 cm
Width (English) 1.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.