Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Double Layer Hot Mill Gloves - Band Top

Catalog No. 19013541 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-013541 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-013541
  • $65.90
    $60.65 / Dozen of 12

    Save $5.25

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $572.50
    $526.65 / Case of 10 DZ

    Save $45.85

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for forging and other operations that require heat protection.

  • Constructed of two cotton layers
  • Ideal for forging and other operations that require heat protection

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Nap-out
Color Natural
Latex Free Yes
Material Cotton
Product Type Hot Mill Gloves
Size Universal
Style Clute
For Use With (Application) Forging, and Hot Metal Handling/Castings/Molds
Gender Men's
Glove Cuff Style Band Top
Series 94-924
Yarn Weight 24 oz.
Length (English) 10.3 in.
Length (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.