Learn More
Fisherbrand™ Humidity Sponges™
SCI_ED
Catalog No. S33516IND Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Humidity Sponge
Humidity Sponge, Indicating
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S33516IND Humidity Sponge, Indicating
S33516REG Humidity Sponge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S33516IND
Add to Cart
Add to Cart
Humidity Sponges™ turn any container into a desiccator.
  • Meets all scientific drying requirements
  • Drying capacity depends on volume
  • Indicator Pack contains a blue indicating gel that turns pink when the desiccant pack has reached its moisture capacity and must be replaced.
  • Reusable Pack may be reused repeatedly and does not have a color indicator.
  • Regeneration requires oven heating to 104°C (220°F). Single relative humidity card shows when regeneration is required.
Specifications
Material Humidity Sponge, Indicating
Quantity 40/Pk.
Packaging Dessicant Bag
Weight 10 g