Learn More
Fisherbrand™ Impact Goggles
SCI_ED
Catalog No. S01823 Shop All Fisherbrand Products
Change view
Click to view available options
Impact goggles; PC lens w/antifog, antiscratch coating
Catalog No.
S01823 Impact goggles; PC lens w/antifog, antiscratch coating
1 options
This item is not returnable. View return policy
Catalog No. S01823
This item is not returnable. View return policy
Lens coated to minimize scratching and fogging.
  • Built for comfortable protection with a soft, flexible PVC body that conforms to facial contours
  • Large frames fit over most prescription eyewear
  • Coated, clear polycarbonate lens resists impact, scratching and fogging
  • Headstrap easily adjusts for a custom fit
  • Protect eyes from dust, mist and other airborne particles
  • perforations on goggle top and sides provide direct ventilation
Compliance
Meet ANSI™ Z87.1-2003
Specifications
Certifications/Compliance ANSI Z87.1-2003
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Impact Goggle
Ventilation Details Direct vents on sides and top
Material Polycarbonate