Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Infecon Infectious Substance Mini Label

Catalog No. 19038302b Shop All Fisherbrand Products
Change view
Click to view available options
Size (Metric):
15.2 x 17.7 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
19-038-302B 15.2 x 17.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-038-302B
Only null left
Add to Cart
Add to Cart

Contains the required 6.2 Warning

  • Has convenient fill-in-the-blank format for specimen identification

Specifications

Product Type Infectious Substance Mini Label
For Use With (Application) For specimen identification
Quantity 12/Pk.
Size (English) 6 x 7 in.
Size (Metric) 15.2 x 17.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.