Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lab Automation Pipet Tips: Bulk Pack
GREENER_CHOICE

Catalog No. 02707271 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02-707-271 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-707-271
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

  • Ideal for demanding HTS procedures
  • Provide maximum performance, accuracy, and reproducibility in lab automation procedures
  • Engineered to ensure proper fit and function with several standard and customized automated liquid-handling systems
  • Choose tips in an assortment of volume sizes and styles

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
For Use With (Application) Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Format Rack
Tip Style Automation
Disposable Single-use
For Use With (Equipment) Packard MultiProbe I
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.