Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Intermediate Grade Cowhide Leather Drivers Gloves

Catalog No. 19013357 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-013-357 Large
19-013-359 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-013-357
Add to Cart
Add to Cart
This item is not returnable. View return policy

Top grain cowhide leather construction provides abrasion resistance and dexterity.

  • Ideal for construction, machine operation, utility work, farming, roping, and other general applications
  • Top-grain leather construction for extended wear and comfort
  • Straight thumb design
  • Launderable

Specifications

Color Natural
Grade Intermediate
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Large
Style Clute
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Open
Hem Color Brown
Thumb Style Straight
Length (English) 9.8 in.
Length (Metric) 24.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.