Learn More
Fisherbrand™ Manifold Pump Tubing
Catalog No. 14190109 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
PVC
Catalog No. Material
14-190-109 PVC
1 options
Catalog No. 14-190-109
Color-coded bridges correspond to tubing ID
Wishbone bridges bracket the 6 in. (15cm) center pumping section of the tubing.
Standard PVC
  • For most routine applications
  • Formulated to yield required durometer hardness and resiliency
  • Supplied in 16 in. (41cm) lengths
Specifications
Material PVC